Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stoml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023231.2 | NP_075720.1 | 4, 5 | 361 | ||
GenBank mRNA | AF323178.1 | - | 4, 5 | 381 | ||
AK002793.1 | - | 4, 5 | 358 | |||
BC003425.1 | - | 4, 5 | 361 | 2609584 | ||
BC069941.1 | - | 4, 5 | 360 | 5390357 |