Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164220.1 | NP_001157692.1 | 3 | 447 | ||
NM_023233.3 | NP_075722.1 | 3 | 385 | |||
GenBank mRNA | AF220129.1 | - | 3 | 462 | ||
AK013959.2 | - | 3 | 357 | |||
AK017850.1 | - | 2 | 342 | |||
AK162129.1 | - | 3 | 435 | |||
BC138576.1 | - | 2 | 311 | 8861596 | ||
BC145915.1 | - | 2 | 311 | 40131218 | ||
BM933374.1 | - | 3 | 385 |