Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023332.3 | NP_075821.1 | 5, 6 | 603 | ||
GenBank mRNA | AF158744.1 | - | 5, 6 | 588 | ||
AK005409.1 | - | 5, 6 | 604 | |||
AK005494.1 | - | 5, 6 | 604 | |||
BC064762.1 | - | 5, 6 | 571 | 30013931 |