Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mesdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023403.3 | NP_075892.3 | 2 | 566 | ||
GenBank mRNA | AK008735.1 | - | 2 | 283 | ||
AK031949.1 | - | 2 | 309 | |||
AK076760.1 | - | 2 | 306 | |||
AK076773.1 | - | 2 | 301 | |||
AK172009.1 | - | 1 | 3362 | |||
AK204742.1 | - | 2 | 138 | |||
BC014742.1 | - | 2 | 287 | 4239248 | ||
BC058612.1 | - | 3220 | ||||
BU054434.1 | - | 2 | 566 |