Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164222.1 | NP_001157694.1 | 5 | 897 | ||
NM_023420.2 | NP_075909.1 | 5 | 897 | |||
GenBank mRNA | AF232932.1 | - | 5 | 911 | ||
AF232934.1 | - | 5 | 911 | |||
AK012989.1 | - | 5 | 878 | |||
AK030305.1 | - | 6 | 805 | |||
AK038061.1 | - | 5 | 887 | |||
AK038803.1 | - | 5 | 878 | |||
AK047100.1 | - | 5 | 894 | |||
AK080958.1 | - | 5 | 895 | |||
AK163624.1 | - | 5 | 887 | |||
BC016197.1 | - | 5 | 860 | 4502118 |