Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168333.1 | NP_001161805.1 | 9 | 1146 | ||
NM_023476.3 | NP_075965.2 | 9 | 1287 | |||
GenBank mRNA | AB047402.1 | - | 9 | 1288 | ||
AB050626.1 | - | 9 | 1124 | |||
AK087142.1 | - | 1147 | ||||
BC005738.1 | - | 9 | 1114 | 2650851 | ||
BC018539.1 | - | 9 | 1131 | 3596560 | ||
CT010275.1 | - | 8 | 905 |