Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023479.2 | NP_075968.2 | 4 | 447 | ||
GenBank mRNA | AF308696.2 | - | 4 | 385 | ||
AK156730.1 | - | 4 | 448 | |||
AK172604.1 | - | 3 | 816 | |||
BC023134.1 | - | 4 | 391 | |||
BC048235.1 | - | 4 | 381 | 6493124 |