Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023500.2 | NP_075989.1 | 2 | 562 | ||
GenBank mRNA | AF155511.1 | - | 2 | 497 | ||
AK007734.1 | - | 2 | 562 | |||
AY534248.1 | - | 2 | 542 | |||
BC027097.1 | - | 2 | 590 | |||
BC125618.1 | - | 2 | 439 | 40130133 | ||
BC125620.1 | - | 2 | 439 | 40130135 |