Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023223.2 | NP_075712.2 | 3, 4 | 657 | ||
GenBank mRNA | AB045313.1 | - | 3, 4 | 561 | ||
AF312208.1 | - | 3, 4 | 554 | |||
AK011131.1 | - | 1, 2 | 44 | |||
AK011723.1 | - | 3, 4 | 656 | |||
AK029424.1 | - | 3, 4 | 609 | |||
AK075998.1 | - | 3, 4 | 605 | |||
AK076030.1 | - | 3, 4 | 609 | |||
AK083459.1 | - | 3, 4 | 486 | |||
AK160204.1 | - | 3, 4 | 669 | |||
AK168650.1 | - | 3, 4 | 609 | |||
BC003215.1 | - | 3, 4 | 594 | 3157842 |