Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023229.2 | NP_075718.2 | 8 | 1473 | ||
GenBank mRNA | AF168682.1 | - | 7 | 1368 | ||
AK002552.1 | - | 8 | 1421 | |||
AK003940.1 | - | 5 | 716 | |||
AK038596.1 | - | 8 | 1425 | |||
AK052667.1 | - | 8 | 1418 | |||
AK139374.1 | - | 7 | 2193 | |||
AK151859.1 | - | 8 | 1425 | |||
AK152369.1 | - | 8 | 1474 | |||
AK159751.1 | - | 8 | 1425 | |||
AK164317.1 | - | 1431 | ||||
AK166508.1 | - | 2018 | ||||
AK167346.1 | - | 7 | 2185 | |||
AK186668.1 | - | 1 | 121 | |||
AK187385.1 | - | 1 | 122 | |||
BC013547.1 | - | 8 | 1390 | 4218374 |