Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023232.3 | NP_075721.3 | 6 | 1265 | ||
GenBank mRNA | AF203914.1 | - | 6 | 1230 | ||
AK002887.1 | - | 6 | 1232 | |||
AK077198.1 | - | 6 | 1246 | |||
AK082903.1 | - | 5 | 1356 | |||
AK133194.1 | - | 1245 | ||||
BC054109.1 | - | 6 | 1259 | 30013833 |