Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105.4 | NP_001096.1 | 11 | 2340 | ||
NM_001111067.2 | NP_001104537.1 | 11 | 2159 | |||
XM_005246939.3 | XP_005246996.1 | 11 | 2211 | |||
XM_005246940.3 | XP_005246997.1 | 11 | 2179 | |||
XM_006712825.3 | XP_006712888.1 | 11 | 2292 | |||
XM_011512106.2 | XP_011510408.1 | 12 | 2259 | |||
XM_011512107.2 | XP_011510409.1 | 11 | 2231 | |||
XM_011512108.2 | XP_011510410.1 | 11 | 3638 | |||
GenBank mRNA | BC033867.1 | - | 11 | 2340 | 5209711 | |
BC036748.1 | - | 11 | 2340 | |||
Z22534.1 | - | 10 | 2013 |