Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291047.1 | NP_001277976.1 | 6 | 1606 | ||
NM_001291053.1 | NP_001277982.1 | 4 | 1200 | |||
NM_023249.6 | NP_075738.1 | 5 | 1480 | |||
GenBank mRNA | AF190624.1 | - | 5 | 1398 | ||
AK006137.1 | - | 4 | 1098 | |||
AK014931.1 | - | 5 | 1305 | |||
AK139453.1 | - | 5 | 1388 |