Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023279.2 | NP_075768.1 | 4 | 1536 | ||
GenBank mRNA | AF312873.1 | - | 4 | 1519 | ||
AK012528.1 | - | 4 | 1551 | |||
AK014302.1 | - | 1 | 850 | |||
AK051298.1 | - | 4 | 1548 | |||
AK082942.1 | - | 4 | 1547 | |||
AK149014.1 | - | 4 | 1548 | |||
BC031357.1 | - | 4 | 1536 | 5363050 | ||
BC088749.1 | - | 4 | 1538 | 6307387 |