Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023289.2 | NP_075778.2 | 3 | 612 | ||
GenBank mRNA | AF221099.1 | - | 3 | 655 | ||
AK004142.1 | - | 3 | 613 | |||
AK005432.1 | - | 3 | 613 | |||
AK005463.1 | - | 3 | 613 | |||
AK145847.1 | - | 3 | 613 | |||
BC099452.1 | - | 3 | 600 | 30787372 |