Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023324.2 | NP_075813.2 | 5 | 901 | ||
GenBank mRNA | AF302503.1 | - | 4 | 1123 | ||
AK013388.1 | - | 1 | 16 | |||
AK151660.1 | - | 5 | 902 | |||
AK152900.1 | - | 5 | 902 | |||
AK152997.1 | - | 5 | 901 | |||
AK153372.1 | - | 5 | 902 | |||
AK167525.1 | - | 5 | 902 | |||
BC016515.1 | - | 4 | 500 | 3600130 |