Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023386.5 | NP_075875.3 | 2 | 708 | ||
GenBank mRNA | AJ251364.1 | - | 2 | 703 | ||
AK007477.1 | - | 2 | 686 | |||
AK018021.1 | - | 2 | 701 | |||
AY562228.1 | - | 2 | 638 | |||
BC006590.1 | - | 1 | 286 | |||
BC024872.1 | - | 2 | 666 | 3990099 |