Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023514.4 | NP_076003.3 | 9, 10 | 973 | ||
GenBank mRNA | AJ250193.1 | - | 5, 6 | 442 | ||
AK009082.1 | - | 9, 10 | 923 | |||
AK150985.1 | - | 9, 10 | 930 | |||
BC022587.1 | - | 4, 5 | 411 | |||
BC051936.1 | - | 8, 9 | 666 | |||
BC069980.1 | - | 7, 8 | 567 | |||
BC116437.2 | - | 9, 10 | 923 | 40104493 |