Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023554.2 | NP_076043.2 | 8 | 870 | ||
GenBank mRNA | AB041566.1 | - | 8 | 854 | ||
AK008670.1 | - | 8 | 855 | |||
AK032963.1 | - | 8 | 856 | |||
BC090637.1 | - | 8 | 843 | 30940523 | ||
BC130012.1 | - | 8 | 860 | 40140312 |