Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cse1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023565.3 | NP_076054.1 | 3 | 212 | ||
GenBank mRNA | AF301152.1 | - | 3 | 212 | ||
BC145393.1 | - | 3 | 197 |