Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159351.1 | NP_001152823.1 | 2 | 115 | ||
NM_023585.4 | NP_076074.2 | 2 | 115 | |||
GenBank mRNA | AF303828.1 | - | 1 | 72 | ||
AK017786.1 | - | 2 | 96 | |||
AK019486.1 | - | 2 | 86 | |||
AK028472.1 | - | 2 | 89 | |||
AK028742.1 | - | 2 | 99 | |||
AK031233.1 | - | 2 | 84 | |||
AK033016.1 | - | 2 | 100 | |||
AK146073.1 | - | 2 | 87 | |||
BC029742.1 | - | 2 | 83 | |||
BC044722.1 | - | 2 | 79 | |||
BC058374.1 | - | 2 | 86 | 5715168 | ||
BC083098.1 | - | 2 | 100 | 30460946 | ||
BC094903.1 | - | 2 | 78 | 30081846 | ||
CJ049747.1 | - | 2 | 99 |