Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023624.4 | NP_076113.1 | 1 | 426 | ||
GenBank mRNA | AF255061.2 | - | 1 | 430 | ||
AK004953.1 | - | 1 | 426 | |||
BC141375.1 | - | 1 | 368 | 9055988 | ||
BC141378.1 | - | 1 | 368 | 9055991 |