Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006252.3 | NP_006243.2 | 3 | 374 | ||
XM_017001692.1 | XP_016857181.1 | 3 | 1839 | |||
XM_017001693.1 | XP_016857182.1 | 3 | 459 | |||
XM_017001694.1 | XP_016857183.1 | 4 | 367 | |||
GenBank mRNA | AK314011.1 | - | 3 | 374 | ||
BC069680.1 | - | 3 | 346 | 7262537 | ||
BC069740.1 | - | 3 | 347 | 7262549 | ||
BC069823.1 | - | 3 | 347 | 7262561 | ||
DA710012.1 | - | 3 | 374 | |||
EF056019.1 | - | 3 | 303 | |||
HQ628625.1 | - | 2 | 161 | |||
U06454.1 | - | 3 | 367 |