Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301230.1 | NP_001288159.1 | 3, 4 | 521 | ||
NM_001301232.1 | NP_001288161.1 | 2, 3 | 414 | |||
NM_023635.6 | NP_076124.1 | 2, 3 | 372 | |||
GenBank mRNA | AB046693.1 | - | 1, 2 | 170 | ||
AB120511.1 | - | 1, 2 | 148 | |||
AB232622.1 | - | 1, 2 | 148 | |||
AF304376.1 | - | 2, 3 | 327 | |||
AK010373.1 | - | 2, 3 | 290 | |||
AK030529.1 | - | 2, 3 | 319 | |||
BB857060.1 | - | 2, 3 | 390 | |||
BC008173.1 | - | 3, 4 | 478 | 3710213 | ||
BC009656.1 | - | 2, 3 | 325 | 3590089 | ||
CJ077071.1 | - | 2, 3 | 372 | |||
CV784419.1 | - | 2, 3 | 283 |