Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023671.2 | NP_076160.1 | 3 | 393 | ||
GenBank mRNA | AK011691.1 | - | 3 | 377 | ||
AK011789.1 | - | 3 | 381 | |||
AK031877.1 | - | 3 | 375 | |||
AK078050.1 | - | 3 | 349 | |||
AK133981.1 | - | 3 | 385 | |||
AK151356.1 | - | 3 | 379 | |||
AK151944.1 | - | 3 | 379 | |||
AK169259.1 | - | 3 | 357 | |||
BC005575.1 | - | 3 | 359 | 3967933 | ||
CX236388.1 | - | 3 | 393 |