Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK191484.1 | - | 1 | 147 | ||
AK220177.1 | - | 2 | 396 | |||
BC007473.1 | - | 2 | 186 | 3496252 | ||
BY300006.1 | - | 2 | 229 |