Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023718.3 | NP_076207.1 | 3 | 291 | ||
GenBank mRNA | AF203701.1 | - | 3 | 295 | ||
AF213260.1 | - | 3 | 295 | |||
AK142587.1 | - | 2 | 149 | |||
AK143970.1 | - | 3 | 292 | |||
BC071214.1 | - | 2 | 141 | 5042082 |