Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023732.2 | NP_076221.1 | 1, 2 | 722 | ||
GenBank mRNA | AF045562.1 | - | 1, 2 | 583 | ||
AK004605.1 | - | 1, 2 | 802 | |||
AK040323.1 | - | 1, 2 | 726 | |||
AK150853.1 | - | 1, 2 | 810 | |||
AK151165.1 | - | 1, 2 | 831 | |||
AK152584.1 | - | 1, 2 | 810 | |||
AK164003.1 | - | 1, 2 | 726 | |||
AK168642.1 | - | 1, 2 | 823 | |||
BC006634.1 | - | 1, 2 | 722 | 3710552 |