Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023733.3 | NP_076222.1 | 4 | 349 | ||
GenBank mRNA | AK004567.2 | - | 4 | 350 | ||
AK037973.1 | - | 3 | 801 | |||
BC006593.1 | - | 4 | 337 | 3156501 | ||
BC012308.1 | - | 4 | 364 | 3500167 |