Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023738.4 | NP_076227.1 | 2 | 192 | ||
GenBank mRNA | AK004894.2 | - | 2 | 193 | ||
AK054410.1 | - | 2 | 199 | |||
AK089235.1 | - | 2 | 196 | |||
AK089256.1 | - | 2 | 178 | |||
AK129009.1 | - | 2 | 104 | |||
AK170421.1 | - | 2 | 185 | |||
AK170644.1 | - | 2 | 191 | |||
AK172467.1 | - | 2 | 154 | |||
BC138843.1 | - | 2 | 109 | 8861865 |