Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023740.2 | NP_076229.2 | 4, 5 | 791 | ||
GenBank mRNA | AF176814.1 | - | 4, 5 | 764 | ||
AK005255.1 | - | 2, 3 | 387 | |||
AK151505.1 | - | 4, 5 | 773 | |||
AK168042.1 | - | 4, 5 | 766 | |||
AK183281.1 | - | 1, 2 | 198 | |||
AK186908.1 | - | 1, 2 | 173 | |||
AK188228.1 | - | 1, 2 | 174 | |||
AK205856.1 | - | 1, 2 | 174 | |||
AK211692.1 | - | 1, 2 | 174 | |||
AK214551.1 | - | 1, 2 | 174 | |||
BC010835.1 | - | 4, 5 | 755 | 4239913 |