Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166547.1 | NP_001160019.1 | 2, 3 | 332 | ||
NM_001166548.1 | NP_001160020.1 | 2, 3 | 336 | |||
NM_001166549.1 | NP_001160021.1 | 2, 3 | 440 | |||
NM_023743.2 | NP_076232.2 | 2, 3 | 347 | |||
GenBank mRNA | AB031388.1 | - | 2, 3 | 168 | ||
AK041598.1 | - | 2, 3 | 298 | |||
AK090363.1 | - | 2, 3 | 238 | |||
AK128990.1 | - | 2, 3 | 438 | |||
AK165678.1 | - | 2, 3 | 335 | |||
BC033410.1 | - | 2, 3 | 182 | 5359290 | ||
CJ072393.1 | - | 2, 3 | 339 |