Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023764.3 | NP_076253.1 | 2, 3 | 289 | ||
GenBank mRNA | AJ242971.1 | - | 2, 3 | 293 | ||
AK015062.1 | - | 232 | ||||
AK016482.1 | - | 2, 3 | 290 | |||
AK045422.1 | - | 2, 3 | 273 | |||
AK049263.1 | - | 2, 3 | 311 | |||
AK049394.1 | - | 2, 3 | 235 | |||
AK078391.1 | - | 299 | ||||
AK161312.1 | - | 2, 3 | 276 | |||
AK189903.1 | - | 1, 2 | 63 | |||
BC053515.1 | - | 2, 3 | 276 | |||
BC062139.1 | - | 2, 3 | 270 | 6334946 |