Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025438.2 | NP_001020609.1 | 3 | 676 | ||
NM_001025439.2 | NP_001020610.1 | 3 | 676 | |||
NM_001293663.1 | NP_001280592.1 | 3 | 676 | |||
NM_001293664.1 | NP_001280593.1 | 3 | 676 | |||
NM_001293665.1 | NP_001280594.1 | 3 | 570 | |||
NM_001293666.1 | NP_001280595.1 | 3 | 676 | |||
NM_023813.4 | NP_076302.1 | 3 | 676 | |||
GenBank mRNA | AK012702.1 | - | 3 | 297 | ||
AK020214.1 | - | 3 | 289 | |||
AK032524.1 | - | 3 | 623 | |||
AK039076.1 | - | 3 | 285 | |||
AK042722.1 | - | 3 | 618 | |||
AK044872.1 | - | 3 | 618 | |||
AK142435.1 | - | 3 | 619 | |||
AK145106.1 | - | 3 | 608 | |||
AK148840.1 | - | 3 | 383 | |||
AK157736.1 | - | 3 | 618 | |||
AK220516.1 | - | 3 | 676 | |||
BC042895.1 | - | 3 | 228 | |||
BC052894.1 | - | 3 | 287 | 30055337 | ||
BC057103.1 | - | 3 | 486 |