Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023852.5 | NP_076341.1 | 3 | 458 | ||
GenBank mRNA | AB232589.1 | - | 3 | 270 | ||
AF312037.1 | - | 3 | 270 | |||
AJ310532.1 | - | 3 | 270 | |||
AK012468.1 | - | 3 | 451 | |||
AK014050.1 | - | 3 | 437 | |||
AK014132.1 | - | 3 | 380 | |||
AK079574.1 | - | 3 | 355 | |||
AK148904.1 | - | 3 | 379 | |||
AK161006.1 | - | 3 | 356 | |||
AK164316.1 | - | 3 | 403 | |||
AY026947.1 | - | 3 | 339 | |||
BC114335.2 | - | 3 | 331 | 40045744 | ||
CJ123980.1 | - | 3 | 458 |