Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166024.1 | NP_001159496.1 | 1, 2 | 298 | ||
NM_023854.2 | NP_076343.2 | 1, 2 | 298 | |||
GenBank mRNA | AF229439.1 | - | 1, 2 | 107 | ||
AK009565.1 | - | 1, 2 | 122 | |||
AK149837.1 | - | 1, 2 | 133 | |||
AK168489.1 | - | 1, 2 | 108 | |||
AK169144.1 | - | 1, 2 | 121 | |||
BC005495.1 | - | 1, 2 | 146 | 3487811 | ||
EF437354.1 | - | 1, 2 | 125 |