Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023585.4 | NP_076074.2 | 2, 3 | 215 | ||
GenBank mRNA | AF303828.1 | - | 1, 2 | 172 | ||
AK019486.1 | - | 2, 3 | 186 | |||
AK028472.1 | - | 2, 3 | 189 | |||
AK028742.1 | - | 2, 3 | 199 | |||
AK031233.1 | - | 2, 3 | 184 | |||
AK033016.1 | - | 2, 3 | 200 | |||
AK146073.1 | - | 2, 3 | 187 | |||
BC029742.1 | - | 2, 3 | 183 | |||
BC058374.1 | - | 2, 3 | 186 | 5715168 | ||
BC083098.1 | - | 2, 3 | 200 | 30460946 | ||
BC094903.1 | - | 2, 3 | 178 | 30081846 | ||
BY025619.1 | - | 2, 3 | 223 |