Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023617.2 | NP_076106.2 | 3, 4 | 334 | ||
GenBank mRNA | AF172276.2 | - | 3, 4 | 410 | ||
AK054065.1 | - | 3, 4 | 293 | |||
AK149434.1 | - | 3, 4 | 295 | |||
AY347569.1 | - | 3, 4 | 211 | |||
AY347570.1 | - | 3, 4 | 211 | |||
BC024046.1 | - | 3, 4 | 284 | |||
BC049853.1 | - | 3, 4 | 286 | |||
BC138876.1 | - | 3, 4 | 313 | 8861898 |