Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282684.1 | NP_001269613.1 | 9 | 1370 | ||
NM_001282685.1 | NP_001269614.1 | 7 | 1198 | |||
NM_001282686.1 | NP_001269615.1 | 6 | 1107 | |||
NM_024681.3 | NP_078957.2 | 8 | 1298 | |||
XM_005261741.3 | XP_005261798.2 | 9 | 1590 | |||
XM_005261744.2 | XP_005261801.1 | 7 | 1218 | |||
XM_011530374.2 | XP_011528676.1 | 7 | 1644 | |||
XM_011530377.2 | XP_011528679.1 | 8 | 1281 | |||
GenBank mRNA | AK022304.1 | - | 8 | 1274 | ||
BC025403.1 | - | 8 | 1297 | 4824153 | ||
BC031038.1 | - | 9 | 1353 | 5277700 |