Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023625.4 | NP_076114.2 | 2, 3 | 436 | ||
GenBank mRNA | AF217003.1 | - | 2, 3 | 417 | ||
AK004973.1 | - | 2, 3 | 439 | |||
AK029813.1 | - | 2, 3 | 436 | |||
AK030234.1 | - | 2, 3 | 388 | |||
AK032930.1 | - | 2, 3 | 432 | |||
AK042089.1 | - | 432 | ||||
AK043608.1 | - | 2, 3 | 426 | |||
AK046937.1 | - | 2, 3 | 426 | |||
AK150201.1 | - | 2, 3 | 427 | |||
AK164220.1 | - | 2, 3 | 417 | |||
AK170387.1 | - | 2, 3 | 428 | |||
AK170631.1 | - | 2, 3 | 416 | |||
BC026395.1 | - | 2, 3 | 414 | |||
BC038605.1 | - | 2, 3 | 417 | 4921245 | ||
BC048826.1 | - | 2, 3 | 423 | 4196604 | ||
BC060095.1 | - | 2, 3 | 414 |