Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023631.2 | NP_076120.2 | 3 | 310 | ||
GenBank mRNA | AF233581.1 | - | 3 | 237 | ||
AK158427.1 | - | 3 | 311 | |||
AK160953.1 | - | 3 | 276 | |||
BC117975.1 | - | 3 | 237 | 40098789 | ||
BC117976.1 | - | 3 | 237 | 40098791 |