Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135112.1 | NP_001128584.1 | 3 | 450 | ||
NM_023646.4 | NP_076135.3 | 3 | 450 | |||
GenBank mRNA | AF325535.1 | - | 3 | 491 | ||
AF326358.1 | - | 3 | 491 | |||
AK004575.1 | - | 3 | 400 | |||
AK004910.1 | - | 3 | 402 | |||
AK007852.1 | - | 3 | 391 | |||
AK031250.1 | - | 3 | 407 | |||
AK167511.1 | - | 3 | 412 | |||
AK214348.1 | - | 2 | 183 | |||
AY009320.1 | - | 3 | 491 | |||
BC003920.1 | - | 3 | 383 | |||
BC027240.1 | - | 3 | 412 | 2654866 |