Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehhadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023737.3 | NP_076226.2 | 2, 3 | 240 | ||
GenBank mRNA | AK004867.1 | - | 2, 3 | 211 | ||
BB841865.1 | - | 2, 3 | 240 | |||
BC016899.1 | - | 2, 3 | 214 | 4241677 |