Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023740.2 | NP_076229.2 | 5 | 796 | ||
GenBank mRNA | AF176814.1 | - | 5 | 769 | ||
AK005255.1 | - | 3 | 392 | |||
AK151505.1 | - | 5 | 778 | |||
AK168042.1 | - | 5 | 771 | |||
AK183281.1 | - | 2 | 203 | |||
AK186908.1 | - | 2 | 178 | |||
AK188228.1 | - | 2 | 179 | |||
AK205856.1 | - | 2 | 179 | |||
AK207655.1 | - | 2 | 74 | |||
AK211692.1 | - | 2 | 179 | |||
AK214551.1 | - | 2 | 179 | |||
AK218025.1 | - | 2 | 180 | |||
BC010835.1 | - | 5 | 760 | 4239913 |