Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023773.2 | NP_076262.2 | 1, 2 | 322 | ||
GenBank mRNA | AB041598.2 | - | 1, 2 | 323 | ||
AK018784.1 | - | 1, 2 | 300 | |||
AK135958.1 | - | 1, 2 | 273 | |||
AK145518.1 | - | 1, 2 | 287 | |||
AK158773.1 | - | 1, 2 | 300 | |||
AK159495.1 | - | 1, 2 | 300 | |||
BC079558.1 | - | 1, 2 | 255 | |||
BC145164.1 | - | 1, 2 | 247 | |||
BC145700.1 | - | 1, 2 | 247 | 40131003 | ||
BY111264.1 | - | 1, 2 | 323 |