Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025438.2 | NP_001020609.1 | 4 | 743 | ||
NM_001025439.2 | NP_001020610.1 | 4 | 743 | |||
NM_001293663.1 | NP_001280592.1 | 4 | 743 | |||
NM_001293664.1 | NP_001280593.1 | 4 | 743 | |||
NM_001293665.1 | NP_001280594.1 | 4 | 637 | |||
NM_001293666.1 | NP_001280595.1 | 4 | 743 | |||
NM_023813.4 | NP_076302.1 | 4 | 743 | |||
GenBank mRNA | AK012702.1 | - | 4 | 364 | ||
AK032524.1 | - | 4 | 690 | |||
AK039076.1 | - | 4 | 352 | |||
AK042722.1 | - | 4 | 685 | |||
AK044872.1 | - | 4 | 685 | |||
AK142435.1 | - | 4 | 686 | |||
AK145106.1 | - | 5 | 675 | |||
AK148840.1 | - | 4 | 450 | |||
AK157736.1 | - | 4 | 685 | |||
AK220516.1 | - | 4 | 743 | |||
BC042895.1 | - | 4 | 295 | |||
BC052894.1 | - | 4 | 354 | 30055337 | ||
BC057103.1 | - | 4 | 553 |