Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023852.5 | NP_076341.1 | 3 | 463 | ||
GenBank mRNA | AB232589.1 | - | 3 | 275 | ||
AF312037.1 | - | 3 | 275 | |||
AJ310532.1 | - | 3 | 275 | |||
AK012468.1 | - | 3 | 456 | |||
AK014050.1 | - | 3 | 442 | |||
AK014132.1 | - | 3 | 385 | |||
AK079574.1 | - | 3 | 360 | |||
AK148904.1 | - | 3 | 384 | |||
AK161006.1 | - | 3 | 361 | |||
AK164316.1 | - | 3 | 408 | |||
AY026947.1 | - | 3 | 344 | |||
BC114335.2 | - | 3 | 336 | 40045744 | ||
CJ123980.1 | - | 3 | 463 |