Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023557.3 | NP_076046.1 | 6 | 451 | ||
GenBank mRNA | AK145621.1 | - | 6 | 443 | ||
AK164684.1 | - | 5 | 1095 | |||
AK166620.1 | - | 3 | 353 | |||
BC010808.1 | - | 6 | 449 | 4240658 | ||
BC052652.1 | - | 6 | 449 | 30034507 |