Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040107.1 | NP_001035196.1 | 8 | 1115 | ||
NM_001256413.1 | NP_001243342.1 | 7 | 1075 | |||
NM_032369.3 | NP_115745.2 | 8 | 1106 | |||
XM_005253948.2 | XP_005254005.1 | 8 | 1193 | |||
XM_011538838.1 | XP_011537140.1 | 9 | 1531 | |||
XM_011538839.2 | XP_011537141.1 | 9 | 1265 | |||
XM_011538840.2 | XP_011537142.1 | 9 | 1297 | |||
XM_011538841.2 | XP_011537143.1 | 9 | 1275 | |||
XM_011538842.2 | XP_011537144.1 | 9 | 1255 | |||
XM_011538844.2 | XP_011537146.1 | 9 | 1270 | |||
XM_011538845.1 | XP_011537147.1 | 8 | 1150 | |||
XM_011538846.2 | XP_011537148.1 | 9 | 1224 | |||
XM_011538847.1 | XP_011537149.1 | 8 | 1125 | |||
XM_017020026.1 | XP_016875515.1 | 9 | 1239 | |||
XM_017020027.1 | XP_016875516.1 | 8 | 1124 | |||
GenBank mRNA | AK293543.1 | - | 7 | 1041 | ||
BC007277.1 | - | 8 | 1558 | 3343102 | ||
BC009731.1 | - | 8 | 1057 | 3877292 | ||
BC032672.2 | - | 7 | 1679 | 5577070 |