Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032602.1 | NP_115991.1 | 1 | 727 | ||
GenBank mRNA | AF296766.1 | - | 1 | 727 |